missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Crystal Violet, pure, indicator

Crystal Violet, C25H30ClN3, CAS Number-548-62-9, 90-94-8, Basic Violet 3, C.I. 42555, Gentian Violet, 100g, 13, 756, CHEBI:41688, 407.99, Pure, Danger, GHS H Statement: Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Causes serious eye damage. May cause cancer.

Brand:  Thermo Scientific Chemicals 212120250

Code : 29

Additional Details : CAS Number : 548-62-9 Weight : 0.02500kg Transport : UN number : 3077 Chem class : 9 Pack group : III


 View more versions of this product

Product Code. 10646972

  • £21.40 / 25g
Estimated Shipment: 01-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Crystal Violet
Indicator, Pure
C25H30ClN3
13, 756
Basic Violet 3, C.I. 42555, Gentian Violet
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
407.99
Glass bottle
215.0°C
548-62-9,90-94-8
MFCD00011750
15, 4430
Solubility in water: 16g/L (25°C). Other solubilities: soluble in chloroform, 1 g/10 mL alcohol, 1 g/15 mL glycerin, practically insoluble in ether
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
407.986
CHEBI:41688
Pure
25 g
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Crystal Violet

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Carcinogenicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P273-Avoid release to the environment.

Supplemental information
  • MIXTURE LIST-Contains : C.I. Basic violet 1,Michler's ketone

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only