missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Crystal Violet Solution
Use for convenient Gram staining of bacteria cells. Prolab Diagnostics™ Crystal Violet Solution concentrates are 10 times concentrated to give a ready to use stain on dilution.
Brand: Pro-Lab Diagnostics™ PL7002
View more versions of this product
This item is not returnable.
View return policy
Description
- Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions
- Solutions come in a selection of convenient and economical sizes.
- All stains are manufactured in accordance with all current regulatory requirements
- CE labelled
Specifications
Reagent | |
2L | |
Liquid | |
Dark violet | |
<20°C |
Crystal Violet | |
(EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark | |
For Gram staining | |
Ethanol, C.I. basic violet 3, Methanol | |
Gram Stain |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- crystal violet
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
Hazard Statement
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, C.I. basicviolet 3 withof Michler's ketone,methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction