Learn More
Asecos™ CS-CLASSIC-G One-Door Two-Window Safety Chemical Storage Cabinets
For the safe and proper storage of non-flammable chemicals and toxic substances in working areas.
Brand:  Asecos™ 3056304030579
Product Code. 15711008
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Robust construction and longevity – sturdy all-welded construction for high industrial requirements
  • High-quality, scratch-resistant outer body
  • Double-walled door construction
  • In accordance with TRGS 510
  • Available with double or single wing doors with fixed windows
  • Modern bicolor design – body in light gray, wing doors in a variety of colors
  • Ventilation – ready for connection to a technical exhaust system (DN 75), natural ventilation at the cabinet bottom
  • Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
  • Interior equipment available includes pull-out shelves and bottom collecting sumps
  • Powder-coated textured surface
  • No unauthorized use – doors lockable with cylinder locking, integrated into the turning handle
Specifications
Specifications
4 x 30 kg/11 L powder-coated sheet steel pull-out shelves with PP sump
520 x 545 x 1950 mm
Storage of non-flammable chemicals and toxic substances
1
105.5 kg
Steel
493 x 486 x 1835 mm
Wing, 2 windows
Pull-out shelves: 30 kg/11 L each, Maximum load: 600 kg
Powder-coated
Yellow
Safety storage cabinet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Asecos™ CS-CLASSIC-G One-Door Two-Window Safety Chemical Storage Cabinets > Yellow; 4x 11 L pull-out shelves with PP sump