Learn More
BD Biosciences™ CS&T IVD Beads (CE/IVD)
Used to set up your cytometer, perform daily quality control, and determine lyse/wash application settings.
£235.00 - £440.00
Specifications
Product Type | Fluorescent Polystyrene Beads |
---|---|
Stability | The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 8 hours at 2°C–25°CC |
Product Code | Brand | No. of Tests | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Tests | Price | Quantity & Availability | |||||
15846339
|
BD Biosciences™
656046 |
50 Tests |
£235.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15856339
|
BD Biosciences™
656047 |
150 Tests |
£440.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Intended for in vitro diagnostic use on BD FACSCanto™ II flow cytometers running BD FACSDiva™ software.
- Used to set up the cytometer, to perform daily performance quality control (QC), and to determine lyse/wash (LW) application settings.
- Dyed with fluorochromes that are excited by the cytometer's lasers.
Specifications
Fluorescent Polystyrene Beads | |
The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 8 hours at 2°C–25°CC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BD Biosciences™ CS&T IVD Beads (CE/IVD)