missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSM Media for Mav203 Yeast Cells
A specifically designed media for making agar plates that enable the growth of Saccharomyces cerevisae MaV203 cells
Brand: Invitrogen™ A13292
Code : 50
Additional Details : Weight : 0.44000kg
Description
The CSM Media for Mav203 Yeast Cells is a specifically designed media for making agar plates that enable the growth of Saccharomyces cerevisae MaV203 cells. This product comes with enough reagents to prepare 2 liters of solid media with the following components per liter:- CSM (without tryptophan): 0.74g
- Yeast Nitrogen Base: 6.66g
- Ammonium Sulfate: 5g
- Glucose: 20g
- Agar: 20g
Compatible with All MaV203 Applications
The CSM Media is optimized for the GeneArt™ High-Order Genetic Assembly System, including the recovery for transformations of vectors once they have been adapted to yeast using the GeneArt™ High-Order Vector Conversion Cassette. The media can also be used to grow MaV203 competent yeast cells, both library and sub-cloning scale (cat# 11281-011 and 11445-012).
Plate Preparation Protocol
For each liter, use one pouch of CSM -Trp Agar Yeast Media minus Glucose and 1 bottle of 20% Glucose. Dissolve the contents of the pouch in 900mL of distilled water and autoclave for 20 minutes on liquid cycle. Allow the solution to cool to 55°C and then add one bottle of 20% Glucose (100mL). Mix well and pour plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
CSM Media for MaV203 Yeast Cells has enough reagents to prepare 2 liters of agar media. The product contains two pouches of premixed powder and two 100 ml bottles of filter-sterilized, 20% glucose. Store all components at room temperature. All components are guaranteed stable for 6 months when properly stored. |
|
CSM Media | |
Microbiology Media | |
S. cerevisiae | |
Optimized for use with the GENEART™ High-Order Genetic Assembly System, A13285 | |
Room Temperature |
Liquid, Powder | |
Autoclave | |
Non-sterile | |
Agar Plates | |
GeneArt™ | |
MaV203 |
For Research Use Only. Not for use in diagnostic procedures.