missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CSM-URA, Powdered Media
Single dropout of Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media
£87.77 - £579.82
Specifications
Certifications/Compliance | Molecular biology Certified™ |
---|---|
Content And Storage | 15°C to 30°C |
Form | Powder |
Product Type | Yeast Media |
Formulation | Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387899
|
MP Biomedicals™
114511222 |
100 g |
£579.82
100g |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
11377899
|
MP Biomedicals™
114511212 |
10 g |
£87.77
10g |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
Description
- Add 0.77g of CSM-Ura to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined single drop-out medium for S.cerevisiae
Applications:
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Molecular biology Certified™ | |
Powder | |
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L |
15°C to 30°C | |
Yeast Media | |
For growth of Saccharomyces cerevisiae |