missing translation for 'onlineSavingsMsg'
Learn More

CSN1S1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen CSN1S1
Concentration 400 μg/mL
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16852025
View Documents
Proteintech
22648-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16842025
View Documents
Proteintech
22648-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CSN1S1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CSN1S1. This antibody recognizes human antigen. The CSN1S1 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

CSN1S1
Western Blot, ELISA
Unconjugated
Human
IgG
400 μg/mL
Polyclonal
Rabbit
1446
Primary
Videos
SDS
Documents

Documents

Product Certifications