missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human CSNK1G2 (CK1 gamma 2), His Tag Recombinant Protein
Human CSNK1G2 (CK1 gamma 2), His Tag Recombinant Protein
£435.00 - £6900.00
Specifications
Accession Number | NP_001310 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 52.5 kDa |
Gene Alias | CSNK1G2 (CK1 γ 2) |
KinaseFamily | CK1 Kinase Family |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10475022
|
Thermo Scientific™
PV3990 |
1 mg |
£6900.00
1mg |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11583447
|
Thermo Scientific™
PV3499 |
10 μg |
£435.00
10µg |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- A serine/threonine kinase implicated in a wide range of cellular functions including vesicular trafficking, DNA damage repair, cell cycle progression, and cytokinesis
Specifications
NP_001310 | |
52.5 kDa | |
CK1 Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.04% Triton X-100, 4 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Baculovirus |
Kinase Assay | |
CSNK1G2 (CK1 γ 2) | |
Human | |
His Tag | |
Dry Ice |