Learn More
CleanSpace™ Technology CST Cleaning & Storage Plug
The CleanSpace CST Cleaning and Storage Plug is designed to be used only with the CleanSpace CST ULTRA Power Unit (CST1012).
Brand:  CleanSpace™ Technology CST1024
Product Code. 17855085
  • £68.40 / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to be used only with the CleanSpace CST ULTRA Power Unit (CST1010)
  • With the filter removed from the CleanSpace CST ULTRA Power Unit, this accessory is used to seal all openings (both bellows and filter air inlet).
  • During cleaning, all openings must be sealed.
  • It is also recommended that all openings are sealed and after cleaning and decontamination, and during storage.
Specifications
Specifications
Cleaning and Storage Plug