Learn More
CleanSpace™ Technology CST Full Face Mask
A CST Full Face Mask is designed for use with the CleanSpace PRO Power Unit (CST1002) and CleanSpace ULTRA Power Unit (CST1012).
Brand:  CleanSpace™ Technology CST1018
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17830705
  • £284.00 / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A CleanSpace CST Full Face Mask clips to the power unit and provides respiratory, face and eye protection.
  • It is comprised of an outer visor, inner mask and a five-point harness to evenly support the mask and power unit and provide ease of movement.
  • Compatible with the CleanSpace PRO Power Unit (CST1002) and CleanSpace ULTRA Power Unit (CST1012).
Specifications
Specifications
500 full cycles
Clear
Full Face Mask
1 x Full Face Mask
un3481
None
Large