Learn More
CleanSpace™ Technology CST Half Mask with Head Harness
A CleanSpace CST Half Masks (including a CleanSpace CST Head Harness) is designed for use with the CleanSpace PRO Power Unit (CST1000) and CleanSpace ULTRA Power Unit (CST 1010).
Brand: CleanSpace™ Technology CST1034
This item is not returnable.
View return policy
Description
- Designed for use with the CleanSpace PRO Power Unit (CST1000) and CleanSpace ULTRA Power Unit (CST 1010).
- Made of soft silicone that readily moulds to the face and does not cause allergic dermatitis.
- The easy-to-locate clips secure the CleanSpace CST Half Mask to the Power Unit.
- Attaches easily to connection points on the mask arms and can be removed for cleaning.
- A CleanSpace CST Half Mask (including a CleanSpace CST Head Harness) is NOT certified intrinsically safe.
Specifications
Black | |
Silicon | |
Small |
Reusable | |
1/2 Mask and Harness |