Learn More
CleanSpace™ Technology CST Harness
The CST Head Harness is specifically designed to aid in supporting the Power Unit for optimal comfort.
Brand: CleanSpace™ Technology CST1021
Additional Details : Weight : 0.09000kg
Description
- Specifically designed to provide additional positioning options of the power unit for optimal wearer comfort.
- Compatible with the CleanSpace PRO Power Unit (CST1002) and CleanSpace ULTRA Power Unit (CST1012).
- Made of light weight polypropylene plastic.
- Clips directly into the Power Unit and is easily changed.
- A CleanSpace CST Neck Support is NOT certified intrinsically safe.
Specifications
Black | |
HDPE, Nylon, High Strength Silicone | |
One Size |
Reusable | |
Harness |