missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSTF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15537-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5′ UTR, but encoding the same protein, have been found for this gene.Specifications
CSTF1 | |
Polyclonal | |
Unconjugated | |
CSTF1 | |
CSTF1 | |
CSTF1 Fusion Protein Ag7874 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q05048 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1477 | |
-20°C | |
Liquid |