Learn More
Thermo Scientific™ CTC Instruments Fixed GT Needle Syringe, 500μL
Thermo Scientific™ CTC Instruments Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
Brand: Thermo Scientific™ 365JL720
Code : A0
Additional Details : Weight : 0.20000kg
Description
- Borosilicate glass with metal flanges
- Needles and plungers are stainless steel
- Field-proven syringe design
Specifications
22 G | |
Fixed Needle Syringe | |
0.41 | |
500 μL |