missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTDSPL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17532-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against a region of human CTDSPL2.
Probable phosphatase.Specifications
CTDSPL2 | |
Polyclonal | |
Unconjugated | |
CTDSPL2 | |
CTDSP like 2, CTDSPL2, HSPC058, HSPC129 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
311368, 329506, 51496 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q05D32, Q5XIK8, Q8BG15 | |
Ctdspl2 | |
CTDSPL2 Fusion Protein Ag11581 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction