Learn More
Gibco™ CTS™ AIM V™ SFM
Gibco CTS AIM V SFM (Therapeutic Grade) is the first commercially available, defined, serum-free medium (SFM) for the proliferation and/or manipulation of T cells and dendritic cells manufactured in compliance with cGMP.
Brand: Gibco™ 0870112BK
1231.20 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 11.27100kg
Description
Gibco CTS AIM V SFM (Therapeutic Grade) is the first commercially available, defined, serum-free medium (SFM) for the proliferation and/or manipulation of T cells and dendritic cells manufactured in compliance with cGMP. It contains L-glutamine, 50 μg/mL streptomycin sulfate, and 10 μg/mL gentamicin sulfate.Specifications
Store at 2–8°C and protect from light. | |
Animal Origin | |
Cell Culture Medium | |
Sterile-filtered | |
Cell and Gene Therapy Research, Development, and Manufacturing | |
10 L |
Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes | |
Liquid | |
Serum-free | |
BPC | |
ISO 13485, MDSAP, FDA-registered, 21 CFR 820 | |
Ambient |
For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.