missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ AIM V™ SFM

Product Code. p-7182845
Click to view available options
Quantity:
1 L
10 L
Unit Size:
10 L
1000mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 12083609

Brand: Gibco™ 0870112BK

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

CTS AIM V SFM is a defined, serum-free medium (SFM) for the proliferation and/or manipulation of T cells, dendritic cells, and other immune cells manufactured in compliance with cGMP.

Gibco Cell Therapy Systems (CTS) AIM-V SFM is a defined, serum-free medium specifically designed for the proliferation and/or manipulation of T cells, dendritic cells, and other immune cells that is manufactured in compliance with current Good Manufacturing Practice (cGMP) standards.

CTS AIM V SFM contains L-glutamine, 50 μg/mL streptomycin sulfate, and 10 μg/mL gentamicin sulfate.

CTS quality

Gibco CTS products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485-certified. Gibco CTS products follow USP 'ancillary materials for cell, gene, and tissue-engineered products' within the responsibilities applicable to a supplier.*

Accompanied by documentation to support your regulatory filing

Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.

*Other aspects of USP will be the responsibility of the end-user to assess. Thermo Fisher Scientific cannot fulfill USP in regards to application and therapy-specific aspects (e.g., residual assessment and removal of the Ancillary Material).

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes
Classification Animal Origin
Form Liquid
Product Type Cell Culture Medium
Serum Level Serum-free
Sterility Sterile-filtered
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 10 L
Shipping Condition Ambient
Format BPC
Content And Storage Store at 2–8°C and protect from light.
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.