missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CTS™ (Cell Therapy Systems) N-2 Supplement
CTS™ (Cell Therapy Systems) N-2 Supplement
Brand: Gibco™ A1370701
133.95 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Code : 50
Additional Details : Weight : 0.05600kg
Description
CTS N-2 Supplement is a serum-free supplement for the growth and expression of neuroblastomas as well as post-mitotic neurons in primary cultures from both the peripheral nervous system and the central nervous system. It is a chemically defined, 100X concentrate based on Bottenstein's N-1 formulation. CTS N-2 Supplement is intended to be used with CTS Neurobasal Medium.CTS quality
Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.
Formulated xeno-free
The preparation contains no animal-derived components at the primary component level.
Accompanied by documentation to support your regulatory filing
Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.
For established protocols using N-2 Supplement, access our Gibco Neurobiology Protocol Handbook.
View publications using Gibco neurobiology products at our Gibco CellCite.
Specifications
Store in freezer (-5 to -20°C) and protect from light. |
|
Xeno-free | |
Mammalian Cell Culture | |
Cell Culture Supplement | |
Sterile | |
Dry Ice | |
Endotoxin, Performance, Sterility, Mycoplasma |
Neural Cells | |
100 X | |
Liquid | |
Serum-Free | |
CTS™ | |
Xeno-Free |
For Research Use Only. Not for use in diagnostic procedures.