missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CELLstart™ Substrate

Product Code. 10052783
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10052783

Brand: Gibco™ A1014201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

CELLstart™ Substrate

CELLstart Substrate is a defined substrate that supports human embryonic stem cell (hESC) attachment and expansion of undifferentiated colonies in serum-free medium without the need for feeder cells. It can also be used with mesenchymal and neural stem cells and for the attachment of human feeder cells (e.g., foreskin fibroblasts).

  • Defined substrate for stem cell culture
  • cGMP manufactured to help ensure manufacturability and traceability

GIBCO products are of high quality and are supplied with harmonized documentation such as certificates of analysis and certificates of origin.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Stem Cells (Neural), Stem Cells (iPS - Induced Pluripotent Stem), Stem Cells (Embryonic), Stem Cells (Mesenchymal)
Classification Serum-Free
Content And Storage Store at 2°C to 8°C.
Form Liquid
Product Line Gibco
Quantity 2 mL
Endotoxin Level ≤50 EU/mL, ≤ 50 EU/mL
Format Tube(s)
Phenol Red Indicator No Phenol Red
Purity or Quality Grade hESC-Qualified
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.