missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Detachable Dynabeads™ CD3/CD28

Product Code. 30284818
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 30284818

Brand: Gibco™ A56996

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco CTS Detachable Dynabeads CD3/CD28 magnetic beads are intended for ex vivo isolation and activation of T cells in a simultaneous one-step mechanism.

Gibco CTS Detachable Dynabeads CD3/CD28 magnetic beads are intended for ex vivo isolation and activation of T cells in a simultaneous one-step mechanism. When used in combination with the CTS Detachable Dynabeads Release Buffer (Cat. No. A55883), this offering is the first-of-its-kind cGMP cell selection technology to have an active release mechanism for process development, clinical trial, and commercial manufacturing use.

Features of the CTS Detachable Dynabeads CD3/CD28 include:

  • Empower users with process flexibility by allowing active release of beads from target cells at any time
  • Optimize target cell purity, yield, activation profile, and desired phenotype thanks to the active release mechanism that gives users more control
  • Provide an automation-friendly solution by working directly with the CTS DynaCellect Magnetic Separation System
  • Enable consistent performance in light of biological variability within processes
  • Are suitable for both autologous and allogeneic cell therapy manufacturing processes

Coated with camelid-derived single-domain VHH antibodies that target CD3 and CD28 cell surface markers, these Dynabeads magnetic beads provide both the primary and co-stimulatory signals that are required for activation and expansion of T cells. These Detachable Dynabeads are meant for direct use with the CTS Detachable Dynabeads Release Buffer that has an active ingredient, biotin, that competitively displaces a biotin derivative on the surface of the beads. This proprietary mechanism allows users to achieve active release of the beads from target cells at any time within their process.

Based on a recommended 3-to-1 bead-to-target-cell ratio (not total nucleated cell number), one 10-mL vial can theoretically isolate and activate up to 1.33 x 109 T cells.

CTS Detachable Dynabeads CD3/CD28 also include Drug Master Files (DMFs) to support cross-referencing in regulatory submissions prior to clinical trial and commercial manufacturing use.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 4 x 108 beads/mL
Content And Storage CTS™ Detachable Dynabeads™ CD3/CD28, 10 mL

Store at 2°C to 8°C with vial upright to keep beads in suspension.
Color Brown
Material Polystyrene
Product Type Cell Isolation and Activation Reagent
For Use With (Equipment) CTS™ DynaCellect™ Magnetic Separation System
Product Line CTS Detachable Dynabeads
Purity or Quality Grade GMP-grade
Quantity 10 mL
Shipping Condition Ships at 2°C to 8°C with wet (not frozen) gel packs, keep vial upright to keep beads in suspension
Diameter (Metric) 4.5 μm
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.