Learn More
Gibco™ CTS™ DPBS, without calcium chloride, without magnesium chloride
Gibco Cell Therapy Systems (CTS) DPBS is the classic Dulbecco's phosphate-buffered saline. This particular formulation lacks calcium, magnesium, and phenol red.
£48.01 - £261.00
Specifications
Additive | No Calcium, No Magnesium |
---|---|
Classification | AOF |
Content And Storage | Store at 15–30°C. |
Dilution | 1X |
Product Type | DPBS |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
17195144
|
Gibco™
A1285602 |
2 L | Bag |
£261.00
2 L |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10368939
|
Gibco™
A1285601 |
1 L | Bottle |
£48.01
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Gibco Cell Therapy Systems (CTS) DPBS is the classic Dulbecco's phosphate-buffered saline. This particular formulation lacks calcium, magnesium, and phenol red.
CTS DPBS is manufactured in cGMP, ISO-certified facilities to help ensure the quality and consistency for reproducible results every day. Tests include osmolality, pH, stability, and the absence of bacterial/fungal contamination and endotoxin.
Gibco CTS products are high quality and supplied with harmonized documentation such as certificates of analysis, certificates of origin, and access to authorization letters for our Drug Master Files, as appropriate. The CTS product line enables you to reduce your burden when qualifying reagents during your transition from research to clinical applications.
Closed systems compatible
This CTS DPBS is provided in bioprocess container format with a chamber constructed from Aegis 5-14 film. The sterile, weldable DEPH-free PVC tubing with Luer lock and C-Flex™ tubing with MPC quick connector provide versatile connection options for closed system manufacturing processes.
Specifications
No Calcium, No Magnesium | |
Store at 15–30°C. |
|
DPBS | |
Cell and Gene Therapy Research, Development, and Manufacturing | |
Room Temperature | |
Liquid |
AOF | |
1X | |
Dulbecco's Phosphate Buffered Saline | |
Sustainable packaging | |
Sterile-filtered | |
ISO13485, MDSAP, FDA-registered, 21 CFR820 |