Learn More
Gibco™ CTS™ GlutaMAX™-I Supplement
GLUTAMAX I, L-alanyl-L-glutamine, is a dipeptide substitute for L-glutamine and can be used as a direct substitute for L-Glutamine at equimolar concentrations in mammalian and stem cell culture with minimal or no adaptation.
Brand: Gibco™ A1286001
Code : 50
Additional Details : Weight : 0.10500kg
Description
GLUTAMAX I, L-alanyl-L-glutamine, is a dipeptide substitute for L-glutamine and can be used as a direct substitute for L-Glutamine at equimolar concentrations in mammalian and stem cell culture with minimal or no adaptation.
The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.
• Animal Orign Free
• cGMP Manufactured to help ensure manufacturability and traceability
• GIBCO™ GlutaMAX™ CTS™ enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.
GLUTAMAX I, L-alanyl-L-glutamine, is a dipeptide substitute for L-glutamine. GLUTAMAX I can be used as a direct substitute for L-glutamine at equimolar concentrations in mammalian cell cultures with minimal or no adaptation. GLUTAMAX I improves growth efficiency and performance of mammalian cell culture systems. GLUTAMAX I eliminates problems associated with the spontaneous breakdown of Lglutamine during incubation. It is highly soluble in aqueous solution and is heat stable. It is supplied as a 200mM (100X), liquid stock in 0.85% NaCl.
GIBCO™ CTS™ products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.
Specifications
Storage Conditions: Room Temperature | |
Xeno-free | |
Liquid | |
Sterile-filtered | |
Sustainable packaging | |
100 mL |
Mammalian and stem cell culture | |
100X | |
Supplement | |
Cell and Gene Therapy Research, Development and Manufacturing | |
ISO 13485, MDSAP, FDA-registered, 21 CFR 820 | |
Room Temperature |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.