missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Immune Cell Serum Replacement (SR)
GREENER_CHOICE

CTS Immune Cell Serum Replacement (SR) is a defined xeno-free formulation designed to support in vitro expansion of human immune cells when added as a serum substitute to supplement a basal cell culture medium such as Gibco CTS OpTmizer T Cell Expansion SFM or CTS AIM V Medium.

£172.00 - £3305.00

Specifications

Classification Xeno-free
Product Type Serum Replacement
Sterility Sterile-filtered
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Shipping Condition Dry Ice
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15433723
View Documents
Gibco™
A2596101
50 mL
£172.00
50mL
Please to purchase this item. Need a web account? Register with us today!
17767187
View Documents
Gibco™
A4702901
250 mL
£934.00
250mL
Please to purchase this item. Need a web account? Register with us today!
15463723
GREENER_CHOICE
View Documents
Gibco™
A2596102
500 mL
£1440.00
500mL
Please to purchase this item. Need a web account? Register with us today!
17777187
View Documents
Gibco™
A4702902
1 L
£3305.00
1 L
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

Xeno-free
Sterile-filtered
Dry Ice
Serum Replacement
ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 18 months from date of manufacture

SDS
Documents

Documents

Product Certifications

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ CTS™ Immune Cell Serum Replacement (SR)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.