missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ KnockOut™ DMEM

KnockOut D-MEM CTS™ is a basal medium optimized for the growth of undifferentiated of human embryonic and induced pluripotent stem cells.

Brand:  Gibco™ A1286101

Code : 50

Additional Details : Weight : 0.61000kg

Product Code. 10378819

  • £46.54 / Each
In Stock
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

KnockOut D-MEM CTS™ is a basal medium optimized for the growth of undifferentiated of human embryonic and induced pluripotent stem cells. The Gibco™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

• Defined D-MEM for hESC/iPSC stem cell culture.
• Osmolarity optimized for growth of undifferentiated hESC and iPSC.
• Gibco™ KnockOut DMEM CTS™ enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.

KnockOut D-MEM CTS™ is a basal medium optimized for the growth of undifferentiated of human embryonic and induced pluripotent stem cells. The osmolarity is optimized to approximate that of mouse embryonic tissue. Contains no L-Glutamine.

Gibco™ CTS™ products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The Gibco™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

Specifications

Specifications

Store at 2–8°C and protect from light.
Animal Origin-free
Cell Culture Medium
Bottle
Sustainable packaging
500 mL
Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
Liquid
Sterile-filtered
Cell and Gene Therapy Research, Development, and Manufacturing
ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Ambient
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers