missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ KnockOut™ SR XenoFree Kit

CTS™ KnockOut™ SR XenoFree Kit

Brand:  Gibco™ A1099202

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10577024

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

GIBCO™ KnockOut™ SR XenoFree CTS™ enables the growth and expansion of human embryonic stem cells (hESC) and human induced pluripotent stem cells (hiPSC) in a cell culture medium containing only human-derived or human recombinant proteins, which facilitates the transition of hESC and hiPSC research from the bench to the clinic.

The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.


  • Defined, xenofree medium for hESC⁄iPSC stem cell culture.
  • First-to-market xenofree hESC⁄iPSC product intended for Ex Vivo tissue and cell culture processing applications.


    GIBCO™ KnockOut™ SR XenoFree CTS™ does not contain bovine or other non-human, animal-derived components. Besides pluripotent stem cell culture expansion and maintenance, KnockOut™ SR XenoFree CTS™ can be used for hESC⁄hiPSC cryopreservation, derivation, and differentiation studies.


    GIBCO™ CTS™ products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate.
    The KnockOut™ SR XenoFree CTS™ Kit includes a bottle of KnockOut™ SR XenoFree CTS™ as a frozen liquid in a 500mL or a 100 mL PET bottle and a product manual.

    This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.
  • Specifications

    Specifications

    One bottle

    Store at -20°C to -5°C. Protect from light.
    Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
    Frozen
    Serum-free
    HSA, Sodium Bicarbonate
    ≤ 10 EU/mL
    cGMP for medical devices, 21 CFR Part 820 and ISO 13485
    CTS™, KnockOut™
    500 mL
    CO2
    Serum-free, Xeno-free
    Stem Cell Media
    Sterile
    No Glucose, No Glutamine, No HEPES, No Phenol Red
    Cell Therapy, Clinical Research, Translational Research
    6 to 8
    ES Cell-Qualified
    Human
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.