Learn More
Gibco™ CTS™ KnockOut™ SR XenoFree Medium

Gibco CTS KnockOut SR XenoFree Medium, when formulated as a complete medium, enables the growth and expansion of human embryonic stem cells (hESC) and human induced pluripotent stem cells (hiPSC) in a cell culture medium containing only human-derived or human recombinant proteins.
£168.00 - £602.00
Specifications
Cell Type | Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs) |
---|---|
Classification | Xeno-free |
Form | Liquid |
Product Type | Cell Culture Supplement |
Sterility | Sterile-filtered |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11519686
![]() |
Gibco™
12618013 |
500 mL |
£602.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11509686
![]() |
Gibco™
12618012 |
100 mL |
£168.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gibco CTS KnockOut SR XenoFree Medium, when formulated as a complete medium, enables the growth and expansion of human embryonic stem cells (hESC) and human induced pluripotent stem cells (hiPSC) in a cell culture medium containing only human-derived or human recombinant proteins, to facilitate the transition of hESC and hiPSC research from the bench to the clinic.
The CTS product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.
CTS KnockOut SR XenoFree Medium does not contain bovine or other non-human, animal-derived components. Besides pluripotent stem cell culture expansion and maintenance, CTS KnockOut SR XenoFree Medium can be used for hESC/hiPSC cryopreservation, derivation, and differentiation studies.
Gibco CTS products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate.
Specifications
Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs) | |
Liquid | |
Sterile-filtered | |
ISO 13485, MDSAP, FDA-registered, 21 CFR 820 | |
Bottle |
Xeno-free | |
Cell Culture Supplement | |
Cell and Gene Therapy Research, Development, and Manufacturing | |
Dry Ice | |
Store at –5 to –20°C and protect from light. |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.