Learn More
Gibco™ CTS™ Neurobasal™-A Medium
CTS Neurobasal-A Medium is designed with the translational stem cell researcher in mind. It can be used to culture post-natal neurons without the need for an astrocyte feeder layer when used with B-27 supplements.
Brand: Gibco™ A1371001
85.49 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.61000kg
Description
CTS Neurobasal-A Medium is designed with the translational stem cell researcher in mind. It can be used to culture post-natal neurons without the need for an astrocyte feeder layer when used with B-27 supplements.
CTS quality
Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.
Accompanied by documentation to support your regulatory filing
Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.
For other applications, we provide alternative formulations of CTS Neurobasal media and B-27 supplements.
For established protocols using Neurobasal media, access our Gibco Neurobiology Protocol Handbook.
View publications using Gibco neurobiology products at our Gibco CellCite.
Specifications
Store at 2–8°C and protect from light. | |
Animal Origin-free | |
Cell Culture Medium | |
Sterile-filtered | |
Cell and Gene Therapy Research, Development, and Manufacturing | |
Ambient |
Neural Cells | |
Liquid | |
Serum-free | |
Bottle | |
ISO 13485, MDSAP, FDA-registered, 21 CFR 820 |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.