missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Neurobasal™ Medium
GREENER_CHOICE

CTS Neurobasal Medium is designed with the translational stem cell researcher in mind. It can be used to culture pre-natal neurons without the need for an astrocyte feeder layer when used with B-27 supplements.

Brand:  Gibco™ A1371201

Product Code. 12043479

  • £97.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

CTS Neurobasal Medium is designed with the translational stem cell researcher in mind. It can be used to culture pre-natal neurons without the need for an astrocyte feeder layer when used with B-27 supplements.

CTS quality

Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.

Accompanied by documentation to support your regulatory filing

Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.

For other applications, we provide alternative formulations of CTS Neurobasal media and B-27 supplements.

For established protocols using Neurobasal media, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 2–8°C and protect from light.
Animal Origin-free
Cell Culture Medium
Sterile-filtered
Cell and Gene Therapy Research, Development, and Manufacturing
500 mL
Neural Cells
Liquid
Serum-free
Bottle
ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Ambient
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ CTS™ Neurobasal™ Medium > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.