missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ NK-Xpander™ Medium

Product Code. p-7221875
Click to view available options
Quantity:
500 mL
5 L
10 L
20 L
Format:
BPC
BPC, Bagged Supplement (100 mL)
Bottle
Unit Size:
5 L
500mL
This item is not returnable. View return policy

Product Code. 17528106

Brand: Gibco™ A5019001

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CTS NK-Xpander Medium is specifically formulated for superior expansion of human natural killer (hNK) cells for cell therapy applications.

Gibco CTS NK-Xpander Medium is specifically formulated for superior expansion of human natural killer (hNK) cells for cell therapy applications. When hNK cells are enriched from human peripheral blood mononuclear cells (hPBMCs) and expanded in a feeder-free system using CTS NK-Xpander Medium supplemented with 5% hAB serum and 500U/mL IL-2, ≥90% of the hNK cells generated maintain CD56+/CD16+/CD3- surface marker expression. Furthermore, such hNK cells demonstrate degranulation and killing of K562 cancer cells when expanded in CTS NK-Xpander Medium.

Superior expansion potential

CTS NK-Xpander Medium is capable of producing a high yield of hNK cells without the need for feeder cells, providing robust expansion of enriched hNK cells from hPBMCs from qualified donors at day 14 to 15 versus other medium options. Cells maintain CD56+/CD16+/CD3- surface marker expression and demonstrate cytolytic capabilities when expanded in CTS NK-Xpander Medium.

Manufactured for cell therapy applications

CTS NK-Xpander Medium is manufactured without cytokines and growth factors to allow flexibility with regard to intended application. The medium does not include human or animal-derived components; however, supplementation with hAB serum is recommended for best results.

CTS quality

Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.* Documentation such as specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available to support your regulatory filing.

The products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485-certified.

Closed-system compatibility

CTS NK-Xpander Medium is available in 500-mL-bottle and 5-L-bag formats for convenient process development and closed-system manufacturing scale-up. The 5-L bioprocess container chamber is constructed from Aegis 5-14 film and includes sterile, weldable DEHP-free PVC tubing with Luer lock and C-Flex™ tubing with MPC quick connector providing versatile connection options for your manufacturing process, including compatibility with the CTS Rotea Counterflow Centrifugation System.

*Other aspects of USP <1043> will be the responsibility of the end-user to assess. Thermo Fisher Scientific cannot fulfill USP <1043> in regards to application and therapy-specific aspects (e.g., residual assessment and removal of the AM).

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Natural Killer (NK) cells
Form Liquid
Product Type Cell Culture Medium
Serum Level Serum-free
Sterility Sterile-filtered
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 500 mL
Shipping Condition Basal: Ambient, Supplement: Dry Ice
Format Bottle
Content And Storage • Basal Medium: Store at 2°C to 8°C and protect from light.
• Supplement: Store at -20°C and protect from light.
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue- Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.