Learn More
Gibco™ CTS™ Opti-MEM™ I Medium
Gibco CTS Opti-MEM I Medium is an improved minimal essential medium (MEM) that is recommended for use with the CTS LV-MAX Transfection Kit as a complexation buffer.
£27.15 - £62.00
Specifications
Cell Line | Viral Production Cells |
---|---|
Cell Type | HEK293 |
Classification | Animal Origin-free |
Form | Liquid |
Product Type | Minimum Essential Medium (MEM) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16386572
|
Gibco™
A4124801 |
100 mL |
£27.15
100mL |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16396572
|
Gibco™
A4124802 |
500 mL |
£62.00
500mL |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Gibco CTS Opti-MEM I Medium is an improved minimal essential medium (MEM) that is recommended for use with the CTS LV-MAX Transfection Kit as a complexation buffer. This medium contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. CTS Opti-MEM I uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH. The formulation of CTS Opti-MEM I is identical to research-grade Opti-MEM I Reduced Serum Medium.
CTS Opti-MEM I is xeno-free and contains human transferrin.
CTS Opti-MEM I is manufactured in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to the ISO 13485. Drug Master File and Regulatory Support File available upon request.
Custom configuration and packaging available upon request.
Specifications
Viral Production Cells | |
Animal Origin-free | |
Minimum Essential Medium (MEM) | |
Bottle | |
ISO 13485, MDSAP, FDA-registered, 21 CFR 820 |
HEK293 | |
Liquid | |
Sterile-filtered | |
LV Production | |
Ambient |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.