missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CTS™ OpTmizer™ T Cell Expansion SFM, bottle format
CTS™ OpTmizer™ T Cell Expansion SFM, bottle format
Brand: Gibco™ A1048501
146.30 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : NEW
Additional Details : Weight : 2.35000kg
Description
GIBCO OpTmizer™ CTS™ T-Cell Expansion SFM has been developed for the growth and expansion of human T lymphocytes. OpTmizer™ CTS™ T-Cell Expansion medium is a complete serum-free, xeno-free 1X medium consisting of OpTmizer™ T-Cell Expansion Basal Medium (1 L bottle) and OpTmizer™ T-Cell Expansion Supplement (26 mL), which are mixed together prior to use.With OpTmizer™ T-Cell Expansion SFM, we offer both basic and clinical research communities a new tool to enhance their success in growing human T-cells. It is specially formulated to allow for superior cell growth & viability, less variability and more consistent results, and easier transition from research to clinical applications.
Workflow benefits:
“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”
Specifications
OpTmizer™ T-Cell Expansion Basal Medium (1 x 1000 mL bottle): Store at 2–8°C. Protect from light. OpTmizer™ T-Cell Expansion Supplement (1 x 26 mL): Store in the dark at 2–8°C. |
|
T Cells | |
Suspension Cell Culture | |
Cell Expansion Serum Free Medium (SFM) | |
Sterile | |
No Glutamine | |
No antibiotics | |
CTS™, OpTmizer™ | |
1 bottle |
CO2 | |
Serum-free, Xeno-free | |
Liquid | |
Serum-free | |
HEPES, Phenol Red, Sodium Pyruvate, Sodium Bicarbonate | |
Very Low | |
Calcium, Magnesium | |
Research Grade |