missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemPro™ MSC SFM

StemPro™ MSC SFM is a serum-free medium (SFM) specially formulated for the growth and expansion of human mesenchymal stem cells (MSCs).

£367.00

Specifications

Content And Storage This kit is shipped in two parts with separate storage requirements. These components are not sold individually:
• StemPro™ MSC SFM Basal Medium, 500 mL (ships at room temperature, store in the dark at 2°C to 8°C)
• StemPro™ MSC SFM Supplement, 75 mL (ships on dry ice, store in the dark at -5°C to -20°C)
Culture Environment CO2
Cell Type Mesenchymal Stem Cells
Classification Serum-free
Form Liquid
View More Specs

Products 1
Product Code Brand With Additives Without Additives Price Quantity & Availability  
Product Code Brand With Additives Without Additives Price Quantity & Availability  
10687193
View Documents
Gibco™
A1033201
Low Glucose, Sodium Pyruvate, Sodium Bicarbonate, Adenosine, Deoxyribonucleosides No Glutamine, No Phenol Red
£367.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

StemPro™ MSC SFM is a serum-free medium (SFM) specially formulated for the growth and expansion of human mesenchymal stem cells (MSCs). StemPro™ MSC SFM enables superior human MSC growth and increased consistency compared to classical serum-supplemented medium (DMEM + 10% FBS).

Using StemPro™ MSC SFM, human MSCs can be expanded beyond 5 passages while still maintaining their tri-lineage mesoderm differentiation potential (i.e., ability to differentiate into osteogenic, chondrogenic and adipogenic lineages). Each lot of StemPro™ MSC SFM is qualified using a human MSC performance assay and human MSCs growing in serum-supplemented medium can be transitioned directly into StemPro™ MSC SFM with little or no adaptation required. In addition, StemPro™ MSC SFM facilitates expansion of MSCs directly from primary human bone marrow.

Specifications

Specifications

This kit is shipped in two parts with separate storage requirements. These components are not sold individually:
• StemPro™ MSC SFM Basal Medium, 500 mL (ships at room temperature, store in the dark at 2°C to 8°C)
• StemPro™ MSC SFM Supplement, 75 mL (ships on dry ice, store in the dark at -5°C to -20°C)
Mesenchymal Stem Cells
Liquid
Serum-free
Low (≤10 EU/mL)
Research Grade
Human
CO2
Serum-free
Stem Cell Serum Free Media (SFM)
Low Glucose, Sodium Pyruvate, Sodium Bicarbonate, Adenosine, Deoxyribonucleosides
Calcium, Sodium Phosphate
1 Kit
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.