Learn More
Gibco™ CTS™ TrueCut™ Cas9 Protein
Gibco™ CTS TrueCut Cas9 Protein is our highest performing CRISPR-Cas9 protein, engineered to deliver maximum ribonucleoprotein (RNP)-editing efficiency.
£9140.00 - £17519.00
Specifications
Concentration | 10 mg/mL |
---|---|
For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |
Product Type | Cas9 Protein |
Sterility | Sterile-filtered |
Content And Storage | Store at –20°C. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17747187
|
Gibco™
A45220 |
2.5 mg |
£9140.00
2.50mg |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17757187
|
Gibco™
A45221 |
5 mg |
£17519.00
5mg |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Gibco CTS TrueCut Cas9 Protein is our highest performing CRISPR-Cas9 protein, engineered to deliver maximum ribonucleoprotein (RNP)-editing efficiency. This protein is manufactured in compliance with standards for Ancillary Materials for Cell, Gene and Tissue-Based Products and is subjected to stringent quality specifications. It is provided in a high-concentration and large-scale format for process development and clinical research applications.
Features of CTS TrueCut Cas9 Protein include:
• Consistent high editing efficiency—superior CRISPR-Cas9 editing efficiency in all tested cell lines including primary immune cells (>90% editing in primary T cells)
• Higher stringency manufacturing—manufactured to meet the standards for Ancillary Materials for Cell, Gene, and Tissue-Based Products including USP <1043>, Ph.Eur. 5.2.12, and ISO 20399, following the principles of 21 CFR Part 820 in a FDA-registered manufacturing site
• Aseptic manufacturing—entire process completed using single-use technology
• Extensive safety testing—includes sterility, endotoxin, mycoplasma, residual nucleic acids, and proteins
• Sterile filling—finished product is vialed in a Class A/B (Class 100 and ISO 5 equivalents) cleanroom
• Traceability documentation—COA, COO, Drug Master Files, and Regulatory Support Files are available
CTS TrueCut Cas9 Protein is a Streptococcus pyogenes Cas9 (wt) protein, produced by recombinant DNA technology in E. coli for genome editing with CRISPR technology. Cas9 protein forms a very stable RNP complex with the guide RNA (gRNA) component of the CRISPR/Cas9 system. Incorporation of NLS aids delivery to the nucleus, increasing the editing efficiency and on-target cleavage.
If you would like to evaluate our TrueCut Cas9 protein or conduct small-scale research experiments, we recommend using our TrueCut Cas9 Protein v2 (Cat. No. A36497).
Specifications
10 mg/mL | |
Cas9 Protein | |
Store at –20°C. | |
E. coli | |
Genome Editing |
Cell and Gene Therapy Research, Development, and Manufacturing | |
Sterile-filtered | |
Dry Ice | |
Liquid | |
ISO13485 |
For Research Use or Non-Commercial Manufacturing of Cell-Based Products for Clinical Research. Caution: Not intended for direct administration into humans or animals.