missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ TrypLE™ Select Enzyme
GREENER_CHOICE

Product Code. 12093745
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 12093745

Brand: Gibco™ A1285901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

TrypLE™ Select CTS™, a non-animal alternative for porcine Trypsin, is a recombinant enzyme derived from microbial fermentation.

TrypLE™ Select CTS™, a non-animal alternative for porcine Trypsin, is a recombinant enzyme derived from microbial fermentation. The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

• GIBCO™ TrypLE™ Select ™ CTS™ enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.
• Non-animal or human origin
• Reduced risk of viral or prion contamination
• Removal kinetics and recovery viability comparable to porcine trypsin
• Demonstration of activity on multiple cell lines, including strongly attached lines
• Stable at room temperature (15° to 30°C) for up to 24 months.
• Manufactured using dedicated, autoclaved or disposable equipment.

TrypLE™ Select CTS™, a non-animal alternative for porcine Trypsin, is a recombinant enzyme derived from microbial fermentation. This material is used for the dissociation of attachment-dependent cell lines from plasticware. TrypLE Select has demonstrated the ability to dissociate cells cultured both in serum-free and serum supplemented systems. The product is provided as a 1X stock solution, formulated in DPBS with 1mM EDTA. The concentration of rProtease is proprietary to
Invitrogen Corp.

GIBCO™ CTS™ products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Cell Type Mammalian Cells
Classification Animal Origin-free
Concentration 1X
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile-filtered
Without Additives No Phenol Red
Format Bottle(s)
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Green Features Sustainable packaging
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 100 mL
Shipping Condition Room Temperature
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.