Learn More
Sartorius Cubis™ II High-Capacity Micro Balance, 66S
The Cubis™ II range of high-capacity micro balances with a maximum load between 32 g and 111 g and a readability between 0.001 mg and 0.002 mg provide the ideal choice for a broad range of applications.
Brand:  Sartorius MCA66S-3S00-D HWL
Additional Details : Weight : 14.30000kg
 View more versions of this product
Product Code. 17987390
  • £34703.00 / Each
Estimated Shipment: 12-09-2024
to see stock.
Description
Description
  • Ultra-high resolution with up to 61 million weighing steps (digits).
  • End-to-end data integrity.
  • 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management.
  • Cleanability, part of compliance.
  • Error-free operation.
  • Individual QApp workflows, motorized auto-leveling.
  • Allow upgrades for hardware features like automated draft shields or built-in ionizer.
  • Optional inner draft shield for best weighing performance.
Specifications
Specifications
MCA
Color Touchscreen
66S Weighing Module
3.5 sec.
9.4 in.
11.9 in.
61 g
0.001 mg
MCA66S-3S00-D HWL
Balance
240 mm
302 mm