Learn More
Sartorius Cubis™ II Micro Balance, MCE User Interface
Cubis™ II Micro Balance features fully customizable hardware, software, and connectivity
Brand:  Sartorius MCE10.6S-2S00-M
Additional Details : Weight : 9.30000kg
 View more versions of this product
Product Code. 16437185
  • £34620.00 / Each
Estimated Shipment: 01-10-2024
to see stock.
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Specifications
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Color TFT Touchscreen
Balance, Accessories, Power supply, Manual, Factory test report
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
±5 μg
Micro Balance
340/355 mm weighinh/electrical module
129/61 mm weighing/electrical module
120/240 V
50/60 Hz
10.1 g
1 μg
±3 μg
Draft shield M: Motorized
5 sec.
10 years
139/240 mm
7.2, 7.9 kg
100/240 V ±10%