Learn More
Sartorius Cubis™ II Precision (2-place) Balance, MCE User Interface
GREENER_CHOICEBIOPHARMA
Cubis™ II Precision (2-Place) Balance features fully customizable hardware, software, and connectivity
£4680.00 - £13275.00
Specifications
Connection Type 2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Display Type TFT Touchscreen
Model No draft shield
Type Precision Balance
Warranty 10 years
View More Specs
Products 6
Product Code Brand Capacity Includes Price Quantity & Availability  
Product Code Brand Capacity Includes Price Quantity & Availability  
16477445
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
MCE6202P-2S00-0
1.5 kg, 3 kg, 6.2 kg Balance, Accessories, Power supply, Manual, Factory test report
£4680.00
Each
16223272
BIOPHARMA
View Documents
Sartorius
MCE32202P0-S00
4.2 kg, 32.2 kg Balance, Accessories, Power supply, Manual, Factory test report, QP99 software
£13275.00
1 unit
16203282
BIOPHARMA
View Documents Promotion Details
Sartorius
MCE4202S0-S00
4.2 kg Balance, Accessories, Power supply, Manual, Factory test report, QP99 software
£5135.00
1 unit
16223282
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
MCE6202S-2S00-0
6.2 kg Balance, Accessories, Power supply, Manual, Factory test report, QP99 software
£6020.00
1 unit
16407495
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Sartorius
MCE8202S-2S00-0
8.2 kg Balance, Accessories, Power supply, Manual, Factory test report
£6740.00
Each
16203262
BIOPHARMA
View Documents
Sartorius
MCE2202S0-S00
2.2 kg Balance, Accessories, Power supply, Manual, Factory test report, QP99 software
£4840.00
1 unit
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Warranty
10-year warranty
Specifications
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
No draft shield
10 years
50/60 Hz (± 5%)
TFT Touchscreen
Precision Balance
100/240 V
Class II
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Sartorius Cubis™ II Precision (2-place) Balance, MCE User Interface