missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cultrex 3D Culture Matrix Laminin I
Applications: Bioactivity
Brand: R&D Systems™ 3446-005-01
693.72 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
- Promotes cell attachment and spreading of MG63 human osteosarcoma cells
- Used as a gel or media additive with other basement membrane components
- Helps to study cellular growth and differentiation in three dimensions in vitro
Storage Buffer:
- Dulbecco’s Modified Eagle’s medium containing 10μg/ml gentamycin sulfate and no phenol red
Sterility Testing:
- No bacterial or fungal growth detected after incubation at 37°C for 14 days
- No mycoplasma contamination detected by PCR
Order Info
The product is shipped with dry ice or equivalent. Upon receipt, store it immediately at the temperature recommended below
Specifications
Product is stable for a minimum of 3 months from date of shipment when stored at -80° C in a manual defrost freezer. For optimal stability, store at -80° C in aliquots. Avoid repeated freeze/thaw cycles. | |
Laminin I extracellular matrix protein | |
Bioactivity | |
30 mg |
6 mg/mL | |
<20.0EU per 1μg of the protein by the LAL method | |
≥90% (SDS-PAGE) | |
Murine Engelbreth-Holm-Swarm (EHS) tumor. |
For research use only. Not for use in diagnostic procedures