missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex Mouse Collagen IV

Collagen IV is a major component of basement membranes, which underlies epithelial and endothelial cells and surrounds muscle, fat and nerve cells. It can be used as a thin coating on tissue culture surfaces to promote cell attachment and proliferation and to study its effects on cell behavior.

Brand:  R&D Systems™ 3410-010-02

Product Code. 18789346

  • £184.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

When handling biohazardous materials such as human cells, safe laboratory procedures should be followed and protective clothing should be worn. This product is for research use only and is not approved for diagnostic procedures.

This item is not returnable. View return policy

Description

Description

  • Used as a substrate for cell culture adhesion
  • The recommended working concentration is 1-10 μg/cm2 (1-10 μg/mL) of growth surface depending on cell type
  • Supplied as 2 vials each containing 0.5 mg of protein
  • Source: Murine Engelbreth-Holm-Swarm (EHS) tumor
  • Storage Buffer: 50 mM Hydrochloric Acid, 1.0 mM TCEP
  • Product is stable for 3 months from date of shipment when stored at ≤ -70 °C
  • For optimal stability, store in liquid nitrogen; Avoid freeze-thaw cycles
  • FOR LABORATORY RESEARCH USE ONLY. NOT FOR USE IN DIAGNOSTIC PROCEDURES
Specifications

Specifications

2 vials (0.5 mg/vial). Store at -80 °C in a manual defrost freezer if using the product within 3 months. Avoid repeated freeze-thaw cycles. For optimal stability, store in liquid nitrogen.
1 mg
Murine Engelbreth-Holm-Swarm (EHS) tumor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Results may vary due to variations among tissue/cells derived from different donors or sources