Learn More
R&D Systems™ Cultrex Organoid Harvesting Solution
Depolymerizes basement membrane matrix for harvesting organoids from culture.
Brand: R&D Systems™ 3700-100-01
Additional Details : Weight : 0.00970kg
Description
Embryonic stem (ES) cells are pluripotent stem cells derived from the inner cell mass of pre-implantation embryos. Induced pluripotent stem (iPS) cells can be generated by somatic cell reprogramming following the exogenous expression of specific transcription factors (Oct-3/4, KLF4, SOX2, and c-Myc). These cell types are capable of unlimited, undifferentiated proliferation in vitro and still maintain the capacity to differentiate into a wide variety of somatic cells. In this capacity, pluripotent stem cells have widespread clinical potential for the treatments of heart disease, diabetes, spinal cord injury, and a variety of neurodegenerative disorders.
Applications: Depolymerizes basement membrane matrix for harvesting organoids from culture
Storage: Store unopened product at 2° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Specifications
Store the unopened product at 2 - 8° C. Do not use past expiration date. | |
Organoid Harvesting Solution |
For Research Use Only