missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex Poly-L-Lysine

Applications: Bioactivity

Brand:  R&D Systems™ 3438-100-01

109.72 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18401700

  • £140.00 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Applications: Substrate for cell culture adhesion. An area of 25 cm2 can be coated with 0.5 ml of a 0.01% Poly-L-Lysine solution. Optimal conditions for attachment must be determined for each cell line and application. Slides may be dipped in the solution and air dried before applying sample. Quality Control: Tested for ability to promote attachment of rat PC-12 pheochromocytoma cells. Cell Culture Qualified: No bacterial or fungal growth detected after incubation at 37degreesC for 14 days following USP XXIV, Chapter 71 sterility testing. No mycoplasma contamination detected by PCR.

Order Info

The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.

Specifications

Specifications

Product is stable for at least 6 months from the date of receipt when stored at 2 - 8° C. Keep sterile.
Sterile-filtered
0.01% in Phosphate Buffered Saline, sterile-filtered.
100 mL
Fermentation using starch.
Poly-L-Lysine
<20.0EU per 1μg of the protein by the LAL method
Bioactivity
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only. Not for use in diagnostic procedures