missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex RGF Basement Membrane Extract, Type R1

R&D Systems™ Cultrex Reduced Growth Factor Basement Membrane Type R1 provides a proprietary formulation that has higher tensile strength when compared to other products.

Brand:  R&D Systems™ 3433-001-R1

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18769278

  • £75.59 / 1mL
Estimated Shipment: 29-07-2024
to see stock.

Description

Description

  • Qualified for use in organoid cell culture.
  • Improves take-rate and growth in difficult organoid systems.
  • Rich in Entactin.
  • Quality controlled for peformance consistency.
Specifications

Specifications

Product is stable for a minimum of 3 months from date of shipment when stored at -20°C in a manual defrost freezer. For optimal stability, store (at -80°C. Avoid freeze-thaw cycles.)
8 to 12 mg/mL as determined by Lowry assay
Murine Engelbreth-Holm-Swarm (EHS) tumor
For optimal stability, store at -80°C
1 mL
Multi-Species
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers