Learn More
R&D Systems™ Cultrex RGF Basement Membrane Extract, Type R1
R&D Systems™ Cultrex Reduced Growth Factor Basement Membrane Type R1 provides a proprietary formulation that has higher tensile strength when compared to other products.
Brand:  R&D Systems™ 3433-005-R1
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18776346
  • £248.37 / 5mL
Estimated Shipment: 29-07-2024
to see stock.
Description
Description
  • Avoid freeze-thaw cycles
  • Basement membranes are continuous sheets of specialized extracellular matrix that form an interface between endothelial, epithelial, muscle, or neuronal cells and their adjacent stroma.
  • Basement membranes are degraded and regenerated during development and wound healing.
  • They not only support cells and cell layers, but they also play an essential role in tissue organization that affects cell adhesion, migration, proliferation, and differentiation.
  • Basement membranes provide major barriers to invasion by metastatic tumor cells.
  • Has a higher concentration of entactin, one of the BME components that connects laminins and collagens reinforcing the hydrogel structure.
  • Specifically designed to culture tissue organoids.
Specifications
Specifications
Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.
For optimal stability, store at -80°C
5 mL