Learn More
R&D Systems™ Cultrex Stem Cell Qualified Laminin I
R&D Systems™ Cultrex Stem Cell Qualified Laminin I provides a functionally defined and effective feeder-free surface for the attachment and maintenance of embryonic stem cells in a pluripotent state, thereby enabling its use for growth promotion or study of stem cell differentiation.
Brand: R&D Systems™ 3400-010-03
Additional Details : Weight : 0.00970kg
Specifications
Store at -80°C in a manual defrost freezer | |
1 mg |