Learn More
R&D Systems™ Cultrex UltiMatrix RGF Basement Membrane Extract
Brand: R&D Systems™ BME001-10
Additional Details : Weight : 0.00970kg
Specifications
Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 mL (2 x 5 mL) | |
Mouse |