missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cultrex UltiMatrix RGF Basement Membrane Extract
£75.59 - £532.38
Specifications
Content And Storage | Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
---|---|
Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18779546
|
R&D Systems™
BME001-01 |
1 mL |
£75.59
1mL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18777896
|
R&D Systems™
BME001-05 |
5 mL |
£278.61
5mL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18737206
|
R&D Systems™
BME001-10 |
10 mL (2 x 5 mL) |
£532.38
5mL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
Mouse |