missing translation for 'onlineSavingsMsg'
Learn More

Cumene, 99.9%, extra pure, Thermo Scientific Chemicals

Product Code. 10653132
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-82-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.19
MDL Number MFCD00008881
InChI Key RWGFKTVRMDUZSP-UHFFFAOYSA-N
Synonym isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
PubChem CID 7406
ChEBI CHEBI:34656
IUPAC Name cumene
SMILES CC(C)C1=CC=CC=C1

Specifications

Melting Point -96.0°C
Color Colorless
Density 0.8620g/mL
Boiling Point 152.0°C to 154.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 99.85% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)2
Refractive Index 1.4905 to 1.4925
Quantity 500 mL
Beilstein 05,393
Merck Index 15,2607
Specific Gravity 0.862
Solubility Information Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in alcohol and many other organic solvents,carbon tetrachloride,ether,benzene,acetone
Viscosity 0.79 mPa.s (20°C)
Formula Weight 120.19
Percent Purity 99.9%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Cumene, 99.9%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cumene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P331-Do NOT induce vomiting.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'