missing translation for 'onlineSavingsMsg'
Learn More

Cumene, 99%, pure, Thermo Scientific Chemicals

Catalog Number 10114300
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
98-82-8
C9H12
120.19
MFCD00008881
RWGFKTVRMDUZSP-UHFFFAOYSA-N
isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
7406
CHEBI:34656
cumene
CC(C)C1=CC=CC=C1
This item is not returnable. View return policy
98-82-8
C9H12
120.19
MFCD00008881
RWGFKTVRMDUZSP-UHFFFAOYSA-N
isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
7406
CHEBI:34656
cumene
CC(C)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -96.0°C
Color Colorless
Density 0.8620g/mL
Boiling Point 152.0°C to 154.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)2
Refractive Index 1.4905 to 1.4925
Quantity 2.5 L
Beilstein 05, 393
Merck Index 15, 2607
Specific Gravity 0.862
Solubility Information Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in alcohol and many other organic solvents,carbon tetrachloride,ether,benzene,acetone
Viscosity 0.79 mPa.s (20°C)
Formula Weight 120.19
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Cumene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'