missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupri-Tartaric Solution 2 According to European Pharmacopoeia (EP), Reagecon
High-purity, stable product developed and tested for European Pharmacopoeia (EP) applications including use as a reagent in the performance of tests outlined in several EP monographs.
Brand: Reagecon 1023300-B
This item is not returnable.
View return policy
Description
- European Pharmacopoeia Reagent, Solution
- High-purity, stable product developed and tested for European Pharmacopoeia (EP) applications.
- Applications include use as a reagent in the performance of tests outlined in several EP monographs.
- Accurate, stable and reproducible.
- Stability is proven (either opened or unopened) for the shelf life of this product.
Specifications
HDPE Bottle GL45 | |
500 mL | |
EP Testing | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- CUPRI-TARTARIC SOLUTION II
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide