missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupric Chloride Dihydrate, ≥99%, (ACS Reagent Grade), MP Biomedicals
Click to view available options
Quantity:
500 g
Specifications
Quantity | 500 g |
Assay Percent Range | ≥99% |
Formula Weight | 170.5 |
Grade | ACS Reagent |
Chemical Name or Material | Cupric Chloride Dihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'